Assay Details
Target Gene Details
Entrez Gene ID: | 388591 |
Gene Name: | ring finger protein 207 |
Gene Aliases: |
C1orf188 |
Location: |
Chr.1:6205475-6221299 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF207 | NM_207396.2 | NP_997279.2 | ||
| XM_011541439.2 | XP_011539741.1 | |||
| XM_017001259.1 | XP_016856748.1 | |||
| AK128246.1 | 4 | 1492 | BAC87352.1 | |
| BC119780.1 | 3 | 980 | ||
| DB290694.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831425 | Chr.1:6129291 - 6264418 on Build GRCh38 | Loss |
|
| nsv954005 | Chr.1:6203341 - 6211740 on Build GRCh38 | Deletion |
|
| nsv545236 | Chr.1:6161915 - 6262876 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map