Assay Details
Target Gene Details
Entrez Gene ID: | 54857 |
Gene Name: | glycerophosphodiester phosphodiesterase domain containing 2 |
Gene Aliases: |
GDE3, OBDPF |
Location: |
Chr.X:70423031-70433391 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GDPD2 | NM_001171191.1 | NP_001164662.1 | ||
| NM_001171192.1 | NP_001164663.1 | |||
| NM_001171193.1 | NP_001164664.1 | |||
| NM_017711.3 | NP_060181.2 | |||
| XM_011530977.1 | XP_011529279.1 | |||
| XM_017029614.1 | XP_016885103.1 | |||
| AB048363.1 | BAB13350.1 | |||
| AI277430.1 | ||||
| AK000214.1 | BAA91014.1 | |||
| AK097293.1 | ||||
| AK299255.1 | ||||
| AK301025.1 | ||||
| AK310090.1 | 8 | 1836 | ||
| AK316035.1 | ||||
| AY358986.1 | AAQ89345.1 | |||
| BC032009.1 | AAH32009.1 |
Target Gene Details
Entrez Gene ID: | 105373244 |
Gene Name: | uncharacterized LOC105373244 |
Gene Aliases: |
- |
Location: |
Chr.X:70427358-70436800 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105373244 | XM_011531081.2 | XP_011529383.1 | ||
| XM_011531082.2 | XP_011529384.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33082 | Chr.X:70420685 - 71888183 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map