Assay Details
Target Gene Details
Entrez Gene ID: | 1108 |
Gene Name: | chromodomain helicase DNA binding protein 4 |
Gene Aliases: |
CHD-4, Mi-2b, Mi2-BETA |
Location: |
Chr.12:6570082-6607439 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 27 - Exon 27 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509452 | Chr.12:6566935 - 6653627 on Build GRCh38 | Insertion |
|
| nsv826210 | Chr.12:6561863 - 6612310 on Build GRCh38 | Gain |
|
| nsv973982 | Chr.12:6578894 - 6587817 on Build GRCh38 | Duplication |
|
| nsv832322 | Chr.12:6575884 - 6765914 on Build GRCh38 | Loss |
|
| nsv826211 | Chr.12:6578422 - 6592697 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map