Assay Details
Target Gene Details
Entrez Gene ID: | 246 |
Gene Name: | arachidonate 15-lipoxygenase |
Gene Aliases: |
12-LOX, 15-LOX-1, 15LOX-1 |
Location: |
Chr.17:4630919-4641665 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALOX15 | NM_001140.3 | NP_001131.3 | ||
| AK290309.1 | ||||
| AK296792.1 | ||||
| AK296821.1 | ||||
| AK296904.1 | ||||
| AK312557.1 | ||||
| AK316126.1 | ||||
| BC029032.1 | AAH29032.1 | |||
| M23892.1 | AAA36182.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574273 | Chr.17:4585589 - 5460332 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map