Assay Details
Target Gene Details
Entrez Gene ID: | 57446 |
Gene Name: | NDRG family member 3 |
Gene Aliases: |
- |
Location: |
Chr.20:36651766-36746138 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDRG3 | NM_022477.3 | NP_071922.2 | ||
| NM_032013.3 | NP_114402.1 | |||
| NR_038370.1 | ||||
| XM_006723837.3 | XP_006723900.1 | |||
| XM_006723838.3 | XP_006723901.1 | |||
| XM_011528925.2 | XP_011527227.1 | |||
| XM_011528926.2 | XP_011527228.1 | |||
| XM_011528927.2 | XP_011527229.1 | |||
| XM_011528928.2 | XP_011527230.1 | |||
| XM_017027978.1 | XP_016883467.1 | |||
| XM_017027979.1 | XP_016883468.1 | |||
| XM_017027980.1 | XP_016883469.1 | |||
| AA903471.1 | ||||
| AB044943.1 | BAB20067.1 | |||
| AF308609.1 | AAL08807.1 | |||
| AK027665.1 | BAB55277.1 | |||
| AK128068.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575371 | Chr.20:36629297 - 36654563 on Build GRCh38 | Gain |
|
| nsv833967 | Chr.20:36477427 - 36658869 on Build GRCh38 | Loss |
|
| nsv1057192 | Chr.20:36514051 - 36839310 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map