Assay Details
Target Gene Details
Entrez Gene ID: | 151449 |
Gene Name: | growth differentiation factor 7 |
Gene Aliases: |
BMP12 |
Location: |
Chr.2:20666664-20671490 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GDF7 | NM_182828.3 | 2 | 1404 | NP_878248.2 |
| AB158468.1 | 2 | 886 | BAD07014.1 | |
| AF522369.1 | 2 | 1404 | AAP97720.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508828 | Chr.2:20604120 - 20679078 on Build GRCh38 | Insertion |
|
| nsv527901 | Chr.2:20618698 - 20676626 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75182412] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map