Assay Details
Target Gene Details
Entrez Gene ID: | 55884 |
Gene Name: | WD repeat and SOCS box containing 2 |
Gene Aliases: |
SBA2 |
Location: |
Chr.12:118032687-118062430 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WSB2 | NM_001278557.1 | 2 | 178 | NP_001265486.1 |
| NM_001278558.1 | 2 | 278 | NP_001265487.1 | |
| NM_018639.4 | 2 | 278 | NP_061109.1 | |
| XM_017019642.1 | 2 | 180 | XP_016875131.1 | |
| AF163324.1 | 1 | 115 | AAF80478.1 | |
| AF229181.1 | 2 | 217 | AAF71302.1 | |
| AK294230.1 | ||||
| AK295555.1 | 2 | 179 | ||
| AK298513.1 | 2 | 181 | ||
| AK310892.1 | 2 | 156 | ||
| BC015887.2 | 2 | 168 | AAH15887.1 | |
| DA772463.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3581916 | Chr.12:117356518 - 118228514 on Build GRCh38 | Gain |
|
| esv3630882 | Chr.12:118048289 - 118056870 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map