Assay Details
Target Gene Details
Entrez Gene ID: | 5980 |
Gene Name: | REV3 like, DNA directed polymerase zeta catalytic subunit |
Gene Aliases: |
POLZ, REV3 |
Location: |
Chr.6:111299031-111483715 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| REV3L | NM_001286431.1 | NP_001273360.1 | ||
| NM_001286432.1 | NP_001273361.1 | |||
| NM_002912.4 | NP_002903.3 | |||
| AF078695.1 | AAC28460.1 | |||
| AK310949.1 |
Target Gene Details
Entrez Gene ID: | 643749 |
Gene Name: | TRAF3IP2 antisense RNA 1 |
Gene Aliases: |
C6UAS, C6orf3, NCRNA00248, TRAF3IP2-AS2 |
Location: |
Chr.6:111483472-111602295 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRAF3IP2-AS1 | NR_034108.1 | |||
| NR_034109.1 | ||||
| NR_034110.1 | ||||
| NR_034111.1 | ||||
| BX647114.1 |
More Information
Set Membership: |
|

Genomic Map