Assay Details
Target Gene Details
Entrez Gene ID: | 1508 |
Gene Name: | cathepsin B |
Gene Aliases: |
APPS, CPSB |
Location: |
Chr.8:11842524-11868150 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTSB | NM_001317237.1 | 10 | 1741 | NP_001304166.1 |
| NM_001908.4 | 10 | 1772 | NP_001899.1 | |
| NM_147780.3 | 12 | 1934 | NP_680090.1 | |
| NM_147781.3 | 11 | 1891 | NP_680091.1 | |
| NM_147782.3 | 11 | 1860 | NP_680092.1 | |
| NM_147783.3 | 11 | 1846 | NP_680093.1 | |
| XM_006716244.2 | 12 | 1958 | XP_006716307.1 | |
| XM_006716245.2 | 10 | 2062 | XP_006716308.1 | |
| XM_011543812.2 | 12 | 1867 | XP_011542114.1 | |
| XM_017013097.1 | 11 | 1848 | XP_016868586.1 | |
| XM_017013098.1 | 11 | 1848 | XP_016868587.1 | |
| XM_017013099.1 | 12 | 1890 | XP_016868588.1 | |
| XM_017013100.1 | 11 | 1792 | XP_016868589.1 | |
| XM_017013101.1 | 7 | 1315 | XP_016868590.1 | |
| AK075393.1 | 10 | 1748 | ||
| AK092070.1 | 11 | 1783 | ||
| AK097384.1 | 5 | 1702 | ||
| AK128472.1 | 1 | 2365 | ||
| AK130184.1 | 10 | 1709 | ||
| BC010240.1 | 11 | 1789 | AAH10240.1 | |
| BC095408.1 | 10 | 1709 | AAH95408.1 | |
| BX647765.1 | 10 | 1632 | CAI46053.1 | |
| BX647952.1 | 10 | 1712 | ||
| BX648239.1 | 5 | 7483 | ||
| L16510.1 | 11 | 1774 | AAC37547.1 | |
| M14221.1 | 11 | 1791 | AAA52129.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1034132 | Chr.8:11799524 - 12340733 on Build GRCh38 | Gain |
|
| nsv610293 | Chr.8:11841238 - 12072225 on Build GRCh38 | Gain |
|
| esv34042 | Chr.8:11840828 - 11869308 on Build GRCh38 | Loss |
|
| nsv1018042 | Chr.8:11809987 - 12024876 on Build GRCh38 | Gain |
|
| esv3572516 | Chr.8:11844191 - 11853761 on Build GRCh38 | Loss |
|
| nsv1032739 | Chr.8:11786294 - 11858964 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map