Assay Details
Target Gene Details
Entrez Gene ID: | 822 |
Gene Name: | capping actin protein, gelsolin like |
Gene Aliases: |
AFCP, HEL-S-66, MCP |
Location: |
Chr.2:85394748-85418467 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAPG | NM_001256139.1 | 5 | 575 | NP_001243068.1 |
| NM_001256140.1 | 5 | 865 | NP_001243069.1 | |
| NM_001320732.1 | 5 | 551 | NP_001307661.1 | |
| NM_001320733.1 | 5 | 512 | NP_001307662.1 | |
| NM_001320734.1 | 5 | 865 | NP_001307663.1 | |
| NM_001747.3 | 5 | 865 | NP_001738.2 | |
| XM_011533122.1 | 5 | 685 | XP_011531424.1 | |
| XM_011533123.1 | 5 | 581 | XP_011531425.1 | |
| AK130130.1 | 5 | 865 | ||
| AK225374.1 | 5 | 538 | ||
| AK225378.1 | 5 | 538 | ||
| AK300507.1 | 5 | 457 | ||
| AK309290.1 | 4 | 234 | ||
| AK313898.1 | 5 | 519 | ||
| BC000728.2 | 5 | 519 | AAH00728.1 | |
| BC014549.1 | 5 | 521 | AAH14549.1 | |
| BG762114.1 | 5 | 513 | ||
| BG762391.1 | 5 | 551 | ||
| BU172582.1 | 5 | 517 | ||
| FJ224312.1 | 5 | 721 | ||
| M94345.1 | 5 | 520 | AAA59570.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834283 | Chr.2:85382377 - 85528585 on Build GRCh38 | Loss |
|
| nsv834282 | Chr.2:85237271 - 85414189 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map