Assay Details
Target Gene Details
Entrez Gene ID: | 64137 |
Gene Name: | ATP binding cassette subfamily G member 4 |
Gene Aliases: |
WHITE2 |
Location: |
Chr.11:119149040-119162666 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCG4 | NM_001142505.1 | NP_001135977.1 | ||
| NM_022169.4 | NP_071452.2 | |||
| XM_011542952.2 | XP_011541254.1 | |||
| XM_017018121.1 | XP_016873610.1 | |||
| AJ300465.1 | CAC17140.1 | |||
| AJ308237.1 | CAC87131.1 | |||
| AJ308243.1 | ||||
| AK289830.1 | ||||
| AK290035.1 | ||||
| AK308673.1 | ||||
| AL137563.1 | CAB70814.3 | |||
| BC041091.1 | AAH41091.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv556467 | Chr.11:119154221 - 119193848 on Build GRCh38 | Loss |
|
| nsv468879 | Chr.11:119106009 - 119188695 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map