Assay Details
Target Gene Details
Entrez Gene ID: | 1654 |
Gene Name: | DEAD-box helicase 3, X-linked |
Gene Aliases: |
CAP-Rf, DBX, DDX14, DDX3, HLP2, MRX102 |
Location: |
Chr.X:41333308-41364472 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX3X | NM_001356.4 | 1 | 527 | NP_001347.3 |
| NR_126093.1 | 1 | 527 | ||
| XM_017029313.1 | 1 | 527 | XP_016884802.1 | |
| XM_017029314.1 | 1 | 527 | XP_016884803.1 | |
| AF000982.1 | 1 | 437 | AAC51829.1 | |
| AF000983.1 | 1 | 437 | AAC51830.1 | |
| DA232063.1 | 1 | 121 | ||
| DB065845.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33720 | Chr.X:40891487 - 43157518 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map