Assay Details
Target Gene Details
Entrez Gene ID: | 1629 |
Gene Name: | dihydrolipoamide branched chain transacylase E2 |
Gene Aliases: |
BCATE2, BCKAD-E2, BCKADE2, BCOADC-E2, E2, E2B |
Location: |
Chr.1:100186922-100249864 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DBT | NM_001918.3 | 11 | 4308 | NP_001909.3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830804 | Chr.1:100036777 - 100210481 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map