Assay Details
Target Gene Details
Entrez Gene ID: | 10488 |
Gene Name: | cAMP responsive element binding protein 3 |
Gene Aliases: |
LUMAN, LZIP, sLZIP |
Location: |
Chr.9:35732320-35737008 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CREB3 | NM_006368.4 | 1 | 365 | NP_006359.3 |
| AF029674.1 | 1 | 176 | AAB84166.1 | |
| AF211848.1 | 1 | 350 | AAG43528.1 | |
| AK299781.1 | 1 | 70 | ||
| AK313635.1 | 1 | 82 | ||
| BC009402.2 | AAH09402.1 | |||
| BC010158.2 | AAH10158.1 | |||
| BG701230.1 | 1 | 370 | ||
| U88528.1 | 1 | 87 | AAC04325.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs116051456] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map