Assay Details
Target Gene Details
Entrez Gene ID: | 29115 |
Gene Name: | SAP30 binding protein |
Gene Aliases: |
HCNGP, HTRG, HTRP |
Location: |
Chr.17:75667116-75708062 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SAP30BP | NM_001301839.1 | 1 | 287 | NP_001288768.1 |
| NM_001301855.1 | 1 | 287 | NP_001288784.1 | |
| NM_013260.7 | 1 | 287 | NP_037392.1 | |
| NR_126036.1 | 1 | 287 | ||
| NR_126037.1 | 1 | 287 | ||
| XM_011524693.2 | 1 | 75 | XP_011522995.1 | |
| XM_011524694.2 | 1 | 69 | XP_011522996.1 | |
| XM_011524695.2 | 1 | 66 | XP_011522997.1 | |
| XM_017024545.1 | 1 | 92 | XP_016880034.1 | |
| XM_017024547.1 | 1 | 89 | XP_016880036.1 | |
| XM_017024548.1 | 1 | 66 | XP_016880037.1 | |
| XM_017024549.1 | 1 | 66 | XP_016880038.1 | |
| XM_017024550.1 | XP_016880039.1 | |||
| AF087869.1 | AAM10497.1 | |||
| AF119664.1 | AAF17221.1 | |||
| AF450482.1 | 1 | 287 | AAL47177.1 | |
| AK002202.1 | ||||
| AK023157.1 | ||||
| AK095265.1 | 1 | 57 | BAC04514.1 | |
| AK298060.1 | ||||
| AK310177.1 | ||||
| AY082382.1 | AAL92491.1 | |||
| BC007592.1 | AAH07592.1 | |||
| BC013409.2 | AAH13409.2 | |||
| BC030233.1 | AAH30233.1 | |||
| BC063793.1 | AAH63793.1 | |||
| BM807177.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833544 | Chr.17:75595541 - 75771639 on Build GRCh38 | Loss |
|
| nsv470603 | Chr.17:75468213 - 75741366 on Build GRCh38 | Loss |
|
| nsv516133 | Chr.17:75420733 - 75763223 on Build GRCh38 | Loss |
|
| nsv576027 | Chr.17:75249939 - 75750289 on Build GRCh38 | Gain |
|
| nsv833543 | Chr.17:75517604 - 75692749 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map