Assay Details
Target Gene Details
Entrez Gene ID: | 2135 |
Gene Name: | exostosin like glycosyltransferase 2 |
Gene Aliases: |
EXTR2 |
Location: |
Chr.1:100872372-100895411 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EXTL2 | NM_001033025.2 | 5 | 2523 | NP_001028197.1 |
| NM_001261440.1 | 5 | 2520 | NP_001248369.1 | |
| NM_001261441.1 | 6 | 2547 | NP_001248370.1 | |
| NM_001261442.1 | 4 | 2488 | NP_001248371.1 | |
| NM_001439.3 | 5 | 2559 | NP_001430.1 | |
| NR_048570.1 | 6 | 2654 | ||
| XM_005270621.1 | 6 | 2389 | XP_005270678.1 | |
| XM_011540994.2 | 6 | 2815 | XP_011539296.1 | |
| XM_011540995.2 | 7 | 2832 | XP_011539297.1 | |
| XM_011540996.2 | 6 | 2034 | XP_011539298.1 | |
| XM_017000651.1 | 5 | 2160 | XP_016856140.1 | |
| AF000416.1 | 5 | 2192 | AAC02898.1 | |
| AK001450.1 | 1 | 845 | ||
| AK096914.1 | BAC04893.1 | |||
| BC036015.1 | 5 | 2169 | AAH36015.1 | |
| BC045681.1 | 5 | 3342 | AAH45681.1 | |
| BU689972.1 | 1 | 664 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522064 | Chr.1:100715786 - 101074310 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map