Assay Details
Target Gene Details
Entrez Gene ID: | 22941 |
Gene Name: | SH3 and multiple ankyrin repeat domains 2 |
Gene Aliases: |
AUTS17, CORTBP1, CTTNBP1, ProSAP1, SHANK, SPANK-3 |
Location: |
Chr.11:70467856-71252724 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 29 - Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHANK2 | NM_012309.4 | NP_036441.2 | ||
| NM_133266.4 | NP_573573.2 | |||
| NR_110766.1 | ||||
| XM_005277932.3 | XP_005277989.1 | |||
| XM_017017387.1 | XP_016872876.1 | |||
| XM_017017388.1 | XP_016872877.1 | |||
| XM_017017389.1 | XP_016872878.1 | |||
| XM_017017390.1 | XP_016872879.1 | |||
| AB028945.1 | BAA82974.1 | |||
| AB208025.1 | ||||
| AB208026.1 | ||||
| AB208027.1 | ||||
| AK128563.1 | 1 | 697 | ||
| BC114484.1 | AAI14485.1 | |||
| BX648718.1 | 1 | 1770 |
More Information
Set Membership: |
|

Genomic Map