Assay Details
Target Gene Details
Entrez Gene ID: | 8620 |
Gene Name: | neuropeptide FF-amide peptide precursor |
Gene Aliases: |
FMRFAL |
Location: |
Chr.12:53506688-53507638 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPFF | NM_001320296.1 | NP_001307225.1 | ||
| NM_003717.3 | NP_003708.1 | |||
| AF005271.1 | AAB64288.1 | |||
| BC104234.1 | AAI04235.1 | |||
| BC104235.1 | AAI04236.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv983486 | Chr.12:53503642 - 53514027 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map