Assay Details
Target Gene Details
Entrez Gene ID: | 132200 |
Gene Name: | chromosome 3 open reading frame 49 |
Gene Aliases: |
- |
Location: |
Chr.3:63819365-63848636 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C3orf49 | NR_026866.1 |
Target Gene Details
Entrez Gene ID: | 80145 |
Gene Name: | THO complex 7 |
Gene Aliases: |
NIF3L1BP1, fSAP24, hTREX30 |
Location: |
Chr.3:63833870-63864478 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THOC7 | NM_001285387.2 | NP_001272316.1 | ||
| NM_001285404.1 | NP_001272333.1 | |||
| NM_025075.3 | NP_079351.2 | |||
| XM_006713339.3 | XP_006713402.1 | |||
| AF104914.1 | ||||
| AK027098.1 | BAB15656.1 | |||
| AK225110.1 | ||||
| BC020599.2 | AAH20599.2 | |||
| BC065012.1 | AAH65012.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1008751 | Chr.3:63595517 - 63862172 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map