Assay Details
Target Gene Details
Entrez Gene ID: | 84893 |
Gene Name: | F-box protein, helicase, 18 |
Gene Aliases: |
FBH1, Fbx18, hFBH1 |
Location: |
Chr.10:5889572-5937595 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FBXO18 | NM_001258452.1 | 3 | 826 | NP_001245381.1 |
| NM_001258453.1 | 4 | 898 | NP_001245382.1 | |
| NM_032807.4 | 4 | 914 | NP_116196.3 | |
| NM_178150.2 | 3 | 800 | NP_835363.1 | |
| XM_011519748.2 | 5 | 1494 | XP_011518050.1 | |
| XM_017016842.1 | 4 | 2041 | XP_016872331.1 | |
| XM_017016843.1 | 5 | 1425 | XP_016872332.1 | |
| XM_017016844.1 | 3 | 810 | XP_016872333.1 | |
| XM_017016845.1 | 3 | 1167 | XP_016872334.1 | |
| XM_017016846.1 | 2 | 656 | XP_016872335.1 | |
| AF380349.1 | 4 | 914 | AAM73631.1 | |
| AF454502.1 | 3 | 744 | AAP97700.1 | |
| AF456237.1 | 3 | 756 | AAP97705.1 | |
| AK095343.1 | 3 | 774 | BAC04535.1 | |
| AK122753.1 | 3 | 826 | ||
| AK226128.1 | 3 | 1324 | ||
| AK315173.1 | 2 | 516 | ||
| AL832251.1 | 1 | 319 | ||
| BC006430.1 | 1 | 491 | ||
| BC032674.1 | 1 | 252 | AAH32674.2 | |
| BC080629.1 | 1 | 463 | AAH80629.1 | |
| BC110884.1 | 4 | 823 | AAI10885.1 | |
| BC113375.1 | 3 | 747 | ||
| BC113377.1 | 3 | 747 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831775 | Chr.10:5762372 - 5958886 on Build GRCh38 | Loss |
|
| nsv825229 | Chr.10:5844942 - 5925734 on Build GRCh38 | Gain |
|
| esv3891745 | Chr.10:5902452 - 5919135 on Build GRCh38 | Loss |
|
| dgv1e49 | Chr.10:5684858 - 6183224 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map