Assay Details
Target Gene Details
Entrez Gene ID: | 80135 |
Gene Name: | ribosome production factor 1 homolog |
Gene Aliases: |
BXDC5 |
Location: |
Chr.1:84479237-84498350 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPF1 | NM_025065.6 | 7 | 798 | NP_079341.2 |
| AA761809.1 | ||||
| AF322053.1 | 7 | 753 | AAL37236.1 | |
| AK022537.1 | 7 | 769 | BAB14086.1 | |
| AL359584.2 | 5 | 475 | CAH10671.1 | |
| BC016051.1 | 7 | 760 | AAH16051.1 | |
| CR627468.1 | 5 | 475 | CAH10670.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830437 | Chr.1:84371841 - 84535586 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map