Assay Details
Target Gene Details
Entrez Gene ID: | 7203 |
Gene Name: | chaperonin containing TCP1 subunit 3 |
Gene Aliases: |
CCT-gamma, CCTG, PIG48, TCP-1-gamma, TRIC5 |
Location: |
Chr.1:156308961-156338415 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCT3 | NM_001008800.2 | 6 | 769 | NP_001008800.1 |
| NM_005998.4 | 8 | 883 | NP_005989.3 | |
| NR_036564.1 | 9 | 1045 | ||
| NR_036565.1 | 9 | 996 | ||
| AB208882.1 | 8 | 749 | BAD92119.1 | |
| AK126400.1 | 8 | 670 | ||
| AK293477.1 | 6 | 599 | ||
| AK300765.1 | 9 | 877 | ||
| AK308023.1 | 7 | 575 | ||
| AK315923.1 | 7 | 669 | ||
| AK316387.1 | 9 | 788 | ||
| AL518070.3 | 6 | 574 | ||
| AL833197.1 | 8 | 743 | CAI46192.1 | |
| AY524046.1 | 8 | 708 | AAS94255.1 | |
| BC006501.2 | 8 | 715 | ||
| BC008019.1 | 8 | 725 | AAH08019.1 | |
| BG746513.1 | 8 | 697 | ||
| X74801.1 | 8 | 649 | CAA52808.1 |
More Information
Set Membership: |
|

Genomic Map