Assay Details
Target Gene Details
Entrez Gene ID: | 51052 |
Gene Name: | prolactin releasing hormone |
Gene Aliases: |
PRH, PRRP |
Location: |
Chr.2:237566574-237567175 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRLH | NM_015893.1 | NP_056977.1 | ||
| AB015419.1 | BAA29027.1 | |||
| BC069081.1 | AAH69081.1 | |||
| BC069284.1 | AAH69284.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4192n100 | Chr.2:237564221 - 237600869 on Build GRCh38 | Loss |
|
| nsv1010093 | Chr.2:237552028 - 237659033 on Build GRCh38 | Gain |
|
| nsv1000252 | Chr.2:235899999 - 238766417 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map