Assay Details
Target Gene Details
Entrez Gene ID: | 55970 |
Gene Name: | G protein subunit gamma 12 |
Gene Aliases: |
- |
Location: |
Chr.1:67701466-67833779 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNG12 | NM_018841.5 | 4 | 762 | NP_061329.3 |
| XM_017001809.1 | 4 | 929 | XP_016857298.1 | |
| XM_017001810.1 | 5 | 1170 | XP_016857299.1 | |
| XM_017001811.1 | 5 | 1099 | XP_016857300.1 | |
| AK092014.1 | ||||
| AL832431.1 | 4 | 715 | CAH10645.1 | |
| BC005940.1 | 4 | 743 | AAH05940.1 |
More Information
Set Membership: |
|

Genomic Map