Assay Details
Target Gene Details
Entrez Gene ID: | 2067 |
Gene Name: | ERCC excision repair 1, endonuclease non-catalytic subunit |
Gene Aliases: |
COFS4, RAD10, UV20 |
Location: |
Chr.19:45407333-45478866 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERCC1 | NM_001166049.1 | 2 | 197 | NP_001159521.1 |
| NM_001983.3 | 2 | 197 | NP_001974.1 | |
| NM_202001.2 | 1 | 239 | NP_973730.1 | |
| XM_005258634.1 | 2 | 128 | XP_005258691.1 | |
| XM_005258635.2 | 2 | 598 | XP_005258692.1 | |
| XM_005258636.4 | 2 | 189 | XP_005258693.1 | |
| XM_005258637.1 | 2 | 128 | XP_005258694.1 | |
| XM_011526610.2 | 2 | 144 | XP_011524912.1 | |
| XM_017026459.1 | 2 | 125 | XP_016881948.1 | |
| XM_017026460.1 | 2 | 598 | XP_016881949.1 | |
| XM_017026461.1 | 3 | 227 | XP_016881950.1 | |
| XM_017026462.1 | 3 | 192 | XP_016881951.1 | |
| XM_017026463.1 | 2 | 184 | XP_016881952.1 | |
| XM_017026464.1 | 2 | 185 | XP_016881953.1 | |
| XM_017026465.1 | 2 | 598 | XP_016881954.1 | |
| XM_017026466.1 | 3 | 232 | XP_016881955.1 | |
| AB069681.1 | 2 | 117 | BAB62810.1 | |
| AF001925.1 | 2 | 193 | AAC16253.1 | |
| AF433652.1 | 2 | 172 | AAL56574.1 | |
| AK092039.1 | 2 | 151 | ||
| AK314884.1 | 2 | 129 | ||
| BC008930.2 | 2 | 113 | AAH08930.1 | |
| BC052813.1 | 1 | 182 | AAH52813.1 | |
| BM789972.1 | 1 | 221 | ||
| BQ671113.1 | 2 | 197 | ||
| BT019806.1 | 1 | 51 | AAV38609.1 | |
| DB468563.1 | ||||
| M13194.1 | 2 | 193 | AAA52394.1 | |
| M28650.1 | 2 | 193 | AAA35810.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833845 | Chr.19:45308814 - 45457371 on Build GRCh38 | Loss |
|
| nsv833846 | Chr.19:45381005 - 45469089 on Build GRCh38 | Loss |
|
| nsv428365 | Chr.19:45358027 - 45469089 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map