Assay Details
Target Gene Details
Entrez Gene ID: | 9669 |
Gene Name: | eukaryotic translation initiation factor 5B |
Gene Aliases: |
IF2 |
Location: |
Chr.2:99337371-99400265 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF5B | NM_015904.3 | NP_056988.3 | ||
| AB018284.1 | ||||
| AF078035.1 | AAD16006.1 | |||
| AJ006776.1 | CAB44357.1 | |||
| AK308175.1 | ||||
| AK308176.1 | ||||
| BC032639.1 | AAH32639.1 | |||
| BX485839.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524466 | Chr.2:99335623 - 99415089 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map