Assay Details
Target Gene Details
Entrez Gene ID: | 55026 |
Gene Name: | transmembrane protein 255A |
Gene Aliases: |
FAM70A |
Location: |
Chr.X:120250984-120311556 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM255A | XM_017029619.1 | XP_016885108.1 |
Target Gene Details
Entrez Gene ID: | 10009 |
Gene Name: | zinc finger and BTB domain containing 33 |
Gene Aliases: |
ZNF-kaiso, ZNF348 |
Location: |
Chr.X:120250752-120258396 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZBTB33 | NM_001184742.1 | 3 | 1141 | NP_001171671.1 |
| NM_006777.3 | 2 | 1042 | NP_006768.1 | |
| AK312321.1 | 3 | 1083 | ||
| AL833604.1 | 2 | 983 | CAD91170.1 | |
| AL833856.1 | CAD38715.1 | |||
| BC042753.1 | 2 | 1042 | AAH42753.1 | |
| BX538016.1 | 2 | 996 | CAD97963.1 | |
| BX538101.1 | 2 | 969 | CAD98016.1 |
More Information
Set Membership: |
|

Genomic Map