Assay Details
Target Gene Details
Entrez Gene ID: | 6578 |
Gene Name: | solute carrier organic anion transporter family member 2A1 |
Gene Aliases: |
MATR1, OATP2A1, PGT, PHOAR2, SLC21A2 |
Location: |
Chr.3:133932696-134030076 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLCO2A1 | NM_005630.2 | 14 | 2492 | NP_005621.2 |
| XM_017007075.1 | 14 | 2435 | XP_016862564.1 | |
| XM_017007077.1 | 15 | 2480 | XP_016862566.1 | |
| BC041140.1 | 14 | 2321 | AAH41140.2 | |
| U70867.1 | 14 | 2301 | AAC09469.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs72978388] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map