Assay Details
Target Gene Details
Entrez Gene ID: | 284716 |
Gene Name: | ribosomal modification protein rimK like family member A |
Gene Aliases: |
FAM80A, NAAGS, NAAGS-II |
Location: |
Chr.1:42380797-42425813 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RIMKLA | NM_173642.3 | 4 | 690 | NP_775913.2 |
| XM_006710585.3 | XP_006710648.1 | |||
| XM_017001082.1 | 5 | 749 | XP_016856571.1 | |
| BC039737.1 | 4 | 684 | AAH39737.1 |
More Information
Set Membership: |
|

Genomic Map