Assay Details
Target Gene Details
Entrez Gene ID: | 3621 |
Gene Name: | inhibitor of growth family member 1 |
Gene Aliases: |
p24ING1c, p33, p33ING1, p33ING1b, p47, p47ING1a |
Location: |
Chr.13:110712623-110721074 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ING1 | NM_198217.2 | NP_937860.1 | ||
| NM_198218.2 | NP_937861.1 | |||
| NM_198219.2 | 1 | 963 | NP_937862.1 | |
| AB024401.1 | 1 | 526 | BAA82886.1 | |
| AF044076.1 | 1 | 120 | AAC00501.1 | |
| AF078835.1 | 1 | 76 | AAG12174.1 | |
| AF149721.1 | 1 | 525 | AAF37421.1 | |
| AF149722.1 | AAF37422.1 | |||
| AF149723.1 | AAF37423.1 | |||
| AF181850.1 | 1 | 948 | AAF07921.1 | |
| AJ310392.1 | 1 | 81 | CAC38067.1 | |
| AK302353.1 | ||||
| BC093942.1 | 1 | 128 | AAH93942.1 | |
| BC093944.1 | 1 | 128 | AAH93944.1 | |
| BG703874.1 | ||||
| BQ430264.1 | ||||
| DB450867.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1044234 | Chr.13:110490706 - 110718233 on Build GRCh38 | Gain |
|
| nsv1044340 | Chr.13:110581726 - 111040647 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map