Assay Details
Target Gene Details
Entrez Gene ID: | 961 |
Gene Name: | CD47 molecule |
Gene Aliases: |
IAP, MER6, OA3 |
Location: |
Chr.3:108043094-108094200 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD47 | NM_001777.3 | 11 | 3653 | NP_001768.1 |
| NM_198793.2 | 9 | 3595 | NP_942088.1 | |
| XM_005247908.1 | 10 | 3590 | XP_005247965.1 | |
| XM_005247909.1 | 8 | 3533 | XP_005247966.1 | |
| AK096670.1 | 1 | 1064 | ||
| AL832675.1 | 2 | 5310 | ||
| BC037306.1 | 9 | 3532 | AAH37306.1 | |
| BX640889.1 | 1 | 2222 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011158 | Chr.3:107746970 - 108629120 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map