Assay Details
Target Gene Details
| Entrez Gene ID: | 57863 | 
| Gene Name: | cell adhesion molecule 3 | 
| Gene Aliases: | BIgR, IGSF4B, NECL1, Necl-1, TSLL1, synCAM3 | 
| Location: | Chr.1:159171587-159203142 on Build GRCh38 | 
| Assay Gene Location: | Within Exon 9 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| CADM3 | NM_001127173.1 | 8 | 1141 | NP_001120645.1 | 
| NM_021189.3 | 9 | 1280 | NP_067012.1 | |
| AF062733.2 | 9 | 1237 | AAD17540.2 | |
| AF363367.1 | 8 | 1139 | AAM60749.1 | |
| AK001560.1 | 5 | 539 | BAA91756.1 | |
| AK289541.1 | 9 | 1280 | ||
| AY046418.1 | 8 | 1044 | AAL02143.1 | |
| AY358332.1 | 8 | 1145 | AAQ88698.1 | |
| BC033819.1 | 8 | 1089 | AAH33819.1 | 
Target Gene Details
| Entrez Gene ID: | 100131825 | 
| Gene Name: | CADM3 antisense RNA 1 | 
| Gene Aliases: | - | 
| Location: | Chr.1:159195985-159202422 on Build GRCh38 | 
| Assay Gene Location: | Overlaps Intron 3 - Exon 3 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| CADM3-AS1 | NR_037870.1 | |||
| AK124069.1 | BAC85765.1 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Exonic  Non-exonic  non-DGV Variation | 
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top