Assay Details
Target Gene Details
Entrez Gene ID: | 6712 |
Gene Name: | spectrin beta, non-erythrocytic 2 |
Gene Aliases: |
GTRAP41, SCA5, SCAR14 |
Location: |
Chr.11:66682496-66729361 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 31 - Exon 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPTBN2 | NM_006946.2 | NP_008877.1 | ||
| XM_005274192.4 | XP_005274249.1 | |||
| XM_005274193.3 | XP_005274250.1 | |||
| XM_006718669.3 | XP_006718732.1 | |||
| XM_006718671.3 | XP_006718734.1 | |||
| XM_017018174.1 | XP_016873663.1 | |||
| XM_017018175.1 | XP_016873664.1 | |||
| XM_017018176.1 | XP_016873665.1 | |||
| XM_017018177.1 | XP_016873666.1 | |||
| XM_017018178.1 | XP_016873667.1 | |||
| AB008567.1 | ||||
| BC139788.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518919 | Chr.11:66641938 - 66700998 on Build GRCh38 | Loss |
|
| nsv522277 | Chr.11:66466552 - 66726354 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map