Assay Details
Target Gene Details
Entrez Gene ID: | 9704 |
Gene Name: | DEAH-box helicase 34 |
Gene Aliases: |
DDX34, HRH1 |
Location: |
Chr.19:47349281-47382704 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHX34 | NM_014681.5 | NP_055496.2 | ||
| XM_005259443.3 | XP_005259500.1 | |||
| XM_011527550.2 | XP_011525852.1 | |||
| XM_011527551.2 | XP_011525853.1 | |||
| XM_017027524.1 | XP_016883013.1 | |||
| AK304710.1 | ||||
| D50924.1 | BAA09483.2 | |||
| DR762320.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1062949 | Chr.19:47344392 - 47388294 on Build GRCh38 | Gain |
|
| nsv428366 | Chr.19:47354499 - 47522248 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map