Assay Details
Target Gene Details
Entrez Gene ID: | 7159 |
Gene Name: | tumor protein p53 binding protein 2 |
Gene Aliases: |
53BP2, ASPP2, BBP, P53BP2, PPP1R13A |
Location: |
Chr.1:223779893-223845981 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TP53BP2 | NM_001031685.2 | 12 | 1875 | NP_001026855.2 |
| NM_005426.2 | 13 | 2007 | NP_005417.1 | |
| XM_011544268.1 | 11 | 1736 | XP_011542570.1 | |
| XM_011544269.2 | 9 | 1394 | XP_011542571.1 | |
| XM_017002220.1 | 13 | 2016 | XP_016857709.1 | |
| AJ318888.2 | 12 | 1820 | CAC83012.1 | |
| AK294432.1 | 12 | 1727 | ||
| AK294654.1 | ||||
| AK294964.1 | 5 | 895 | ||
| AK295378.1 | 6 | 599 | ||
| AK316016.1 | 5 | 797 | ||
| AK316103.1 | 5 | 775 | ||
| BC040247.1 | 10 | 1351 | ||
| BC046150.1 | 12 | 1818 | ||
| BC058918.1 | 12 | 1833 | ||
| U58334.1 | 13 | 1952 | AAC50557.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34082 | Chr.1:223663316 - 223956807 on Build GRCh38 | Loss |
|
| esv2759000 | Chr.1:223747518 - 224175461 on Build GRCh38 | Gain+Loss |
|
| nsv832681 | Chr.1:223524706 - 223814989 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map