Assay Details
Target Gene Details
Entrez Gene ID: | 4594 |
Gene Name: | methylmalonyl-CoA mutase |
Gene Aliases: |
MCM |
Location: |
Chr.6:49430360-49463328 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MUT | NM_000255.3 | NP_000246.2 | ||
| XM_005249143.3 | XP_005249200.1 | |||
| AB208845.1 | BAD92082.1 | |||
| AK292568.1 | ||||
| AK312611.1 | ||||
| BC016282.1 | AAH16282.1 | |||
| BT007434.1 | AAP36102.1 | |||
| BX647789.1 | ||||
| KF030882.1 | ||||
| KU178201.1 | ||||
| KU178202.1 | ||||
| M65131.1 | AAA59569.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5960n100 | Chr.6:49322472 - 49684411 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map