Assay Details
Target Gene Details
Entrez Gene ID: | 10243 |
Gene Name: | gephyrin |
Gene Aliases: |
GEPH, GPH, GPHRYN, HKPX1, MOCODC |
Location: |
Chr.14:66507407-67735831 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPHN | NM_001024218.1 | 1 | 194 | NP_001019389.1 |
| NM_020806.4 | 1 | 194 | NP_065857.1 | |
| AB037806.1 | 1 | 194 | ||
| AF272663.1 | 1 | 194 | AAF81785.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525460 | Chr.14:66384467 - 67159877 on Build GRCh38 | Loss |
|
| nsv564953 | Chr.14:65821203 - 66580242 on Build GRCh38 | Gain |
|
| esv2748779 | Chr.14:63663324 - 66595809 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map