Assay Details
Target Gene Details
Entrez Gene ID: | 1001 |
Gene Name: | cadherin 3 |
Gene Aliases: |
CDHP, HJMD, PCAD |
Location: |
Chr.16:68644820-68727573 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDH3 | NM_001317195.1 | NP_001304124.1 | ||
| NM_001317196.1 | NP_001304125.1 | |||
| NM_001793.5 | NP_001784.2 | |||
| XM_011522800.2 | XP_011521102.1 | |||
| AK225396.1 | ||||
| AK296968.1 | ||||
| AK312554.1 | ||||
| BC014462.1 | ||||
| BC041846.1 | AAH41846.1 | |||
| X63629.1 | CAA45177.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833267 | Chr.16:68648596 - 68823901 on Build GRCh38 | Gain |
|
| nsv572908 | Chr.16:68564104 - 68890598 on Build GRCh38 | Gain |
|
| nsv572909 | Chr.16:68607485 - 68678827 on Build GRCh38 | Gain |
|
| esv3638909 | Chr.16:68676046 - 68681679 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map