Assay Details
Target Gene Details
Entrez Gene ID: | 51560 |
Gene Name: | RAB6B, member RAS oncogene family |
Gene Aliases: |
- |
Location: |
Chr.3:133824235-133895847 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB6B | NM_016577.3 | 8 | 3835 | NP_057661.3 |
| XM_011512893.2 | 8 | 3510 | XP_011511195.1 | |
| AB073348.1 | ||||
| AK055102.1 | 1 | 910 | ||
| BC078662.1 | 9 | 3501 | AAH78662.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3569012 | Chr.3:133821680 - 133831917 on Build GRCh38 | Loss |
|
| nsv829729 | Chr.3:133726027 - 133901543 on Build GRCh38 | Loss |
|
| esv3413841 | Chr.3:133824918 - 133826116 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map