Assay Details
Target Gene Details
Entrez Gene ID: | 101927718 |
Gene Name: | DSG1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.18:31343368-31426988 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DSG1-AS1 | NR_110788.1 | |||
| BX476374.1 | ||||
| BX476414.1 |
Target Gene Details
Entrez Gene ID: | 147409 |
Gene Name: | desmoglein 4 |
Gene Aliases: |
CDGF13, CDHF13, HYPT6, LAH |
Location: |
Chr.18:31376777-31415791 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DSG4 | NM_001134453.2 | NP_001127925.1 | ||
| NM_177986.4 | NP_817123.1 | |||
| AY168788.1 | AAO43657.1 | |||
| AY177664.1 | AAO47078.1 | |||
| AY227350.1 | AAP45000.1 | |||
| AY228236.1 | AAP49811.1 | |||
| BC132907.1 | ||||
| BC132909.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3642125 | Chr.18:31388805 - 31392646 on Build GRCh38 | Loss |
|
| dgv611e214 | Chr.18:31380933 - 31436217 on Build GRCh38 | Loss |
|
| dgv3328n100 | Chr.18:31385807 - 31473928 on Build GRCh38 | Gain |
|
| esv3893085 | Chr.18:31384800 - 31429446 on Build GRCh38 | Loss |
|
| dgv975e212 | Chr.18:31385113 - 31471686 on Build GRCh38 | Gain |
|
| esv2751776 | Chr.18:31390184 - 31477756 on Build GRCh38 | Gain |
|
| nsv524890 | Chr.18:31387447 - 31476502 on Build GRCh38 | Gain |
|
| nsv458045 | Chr.18:31389081 - 31476502 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map