Assay Details
Target Gene Details
Entrez Gene ID: | 6130 |
Gene Name: | ribosomal protein L7a |
Gene Aliases: |
L7A, SURF3, TRUP |
Location: |
Chr.9:133348214-133351425 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL7A | NM_000972.2 | NP_000963.1 | ||
| AB056665.1 | BAB39381.1 | |||
| AK222534.1 | ||||
| AK291123.1 | ||||
| AK311743.1 | ||||
| BC023594.2 | AAH23594.1 | |||
| BC023624.2 | AAH23624.1 | |||
| BC071901.1 | AAH71901.1 | |||
| BX641050.1 | 2 | 1916 | ||
| M36072.1 | AAA60282.1 | |||
| X06705.1 | CAA29889.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2739138 | Chr.9:132727483 - 135521553 on Build GRCh38 | Deletion |
|
| nsv972461 | Chr.9:133348790 - 133351286 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map