Assay Details
Target Gene Details
Entrez Gene ID: | 114789 |
Gene Name: | solute carrier family 25 member 25 |
Gene Aliases: |
MCSC, PCSCL, SCAMC-2 |
Location: |
Chr.9:128068200-128109258 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A25 | NM_001006641.3 | NP_001006642.1 | ||
| NM_001006642.3 | NP_001006643.1 | |||
| NM_001265614.2 | NP_001252543.1 | |||
| NM_052901.4 | 1 | 87 | NP_443133.2 | |
| NR_049766.2 | 1 | 87 | ||
| XM_005251688.1 | XP_005251745.1 | |||
| XM_006716948.2 | 1 | 625 | XP_006717011.1 | |
| AB067483.1 | ||||
| AJ619963.1 | CAF04060.1 | |||
| AJ619989.1 | CAF04495.1 | |||
| AJ619990.1 | CAF04496.1 | |||
| AJ619991.1 | CAF04497.1 | |||
| AJ619992.1 | CAF04498.1 | |||
| AK290705.1 | ||||
| AK290991.1 | ||||
| AL559911.3 | ||||
| AY358515.1 | AAQ88879.1 | |||
| BC089448.1 | 1 | 78 | AAH89448.1 | |
| BC103930.1 | AAI03931.1 | |||
| BC103931.1 | AAI03932.1 | |||
| BC103932.1 | AAI03933.1 | |||
| BC103933.1 | AAI03934.1 | |||
| BQ637100.1 | 1 | 87 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1054891 | Chr.9:127939274 - 128102397 on Build GRCh38 | Gain |
|
| nsv615383 | Chr.9:128091848 - 128128781 on Build GRCh38 | Loss |
|
| nsv471324 | Chr.9:128039877 - 128130666 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map