Assay Details
Target Gene Details
Entrez Gene ID: | 56980 |
Gene Name: | PR domain 10 |
Gene Aliases: |
PFM7 |
Location: |
Chr.11:129899706-130003097 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRDM10 | NM_020228.2 | 15 | 2412 | NP_064613.2 |
| NM_199437.1 | 14 | 2400 | NP_955469.1 | |
| NM_199438.1 | 11 | 2138 | NP_955470.1 | |
| NM_199439.1 | 10 | 2126 | NP_955471.1 | |
| XM_011542912.2 | 14 | 2662 | XP_011541214.1 | |
| XM_017018023.1 | 10 | 4211 | XP_016873512.1 | |
| AB033057.2 | 11 | 1922 | BAA86545.2 | |
| AF275817.1 | 11 | 2138 | AAF87243.1 | |
| AF503171.1 | 10 | 2057 | AAP30847.1 | |
| AK000234.1 | 8 | 1367 | BAA91026.1 | |
| AK310354.1 | 15 | 2272 | ||
| AL833172.1 | 14 | 2403 | ||
| BC112934.1 | 14 | 2392 | AAI12935.1 | |
| BC117415.1 | 11 | 1977 | ||
| BC143612.1 | 11 | 1977 | ||
| BC143613.1 | 10 | 1965 | ||
| BX648944.1 | 15 | 2413 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv468893 | Chr.11:129908477 - 129934918 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map