Assay Details
Target Gene Details
Entrez Gene ID: | 118491 |
Gene Name: | cilia and flagella associated protein 70 |
Gene Aliases: |
TTC18 |
Location: |
Chr.10:73253758-73361064 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 27 - Exon 28 |
Target Gene Details
Entrez Gene ID: | 414245 |
Gene Name: | DNAJC9 antisense RNA 1 |
Gene Aliases: |
C10orf103, bA537A6.3 |
Location: |
Chr.10:73247367-73276984 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC9-AS1 | NR_038373.1 | |||
| NR_134458.1 | ||||
| HY014641.1 |
More Information
Set Membership: |
|

Genomic Map