Assay Details
Target Gene Details
Entrez Gene ID: | 7203 |
Gene Name: | chaperonin containing TCP1 subunit 3 |
Gene Aliases: |
CCT-gamma, CCTG, PIG48, TCP-1-gamma, TRIC5 |
Location: |
Chr.1:156308961-156338415 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCT3 | NM_001008800.2 | 9 | 1152 | NP_001008800.1 |
| NM_005998.4 | 11 | 1266 | NP_005989.3 | |
| NR_036564.1 | 12 | 1428 | ||
| NR_036565.1 | 12 | 1379 | ||
| AB208882.1 | 11 | 1132 | BAD92119.1 | |
| AK126400.1 | 11 | 1053 | ||
| AK293477.1 | 9 | 982 | ||
| AK300765.1 | 12 | 1260 | ||
| AK308023.1 | 10 | 958 | ||
| AK315923.1 | 10 | 1052 | ||
| AK316387.1 | 12 | 1171 | ||
| AL833197.1 | 11 | 1126 | CAI46192.1 | |
| AY524046.1 | 11 | 1091 | AAS94255.1 | |
| BC006501.2 | 11 | 1098 | ||
| BC008019.1 | 11 | 1108 | AAH08019.1 | |
| U17104.1 | AAC50068.1 | |||
| X74801.1 | 11 | 1032 | CAA52808.1 |
More Information
Set Membership: |
|

Genomic Map