Assay Details
Target Gene Details
Entrez Gene ID: | 23457 |
Gene Name: | ATP binding cassette subfamily B member 9 |
Gene Aliases: |
EST122234, TAPL |
Location: |
Chr.12:122918708-122975160 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCB9 | NM_001243013.1 | 11 | 2558 | NP_001229942.1 |
| NM_001243014.1 | NP_001229943.1 | |||
| NM_019624.3 | 11 | 2618 | NP_062570.1 | |
| NM_019625.3 | 12 | 2747 | NP_062571.1 | |
| NM_203444.3 | NP_982269.2 | |||
| XM_005253558.2 | 12 | 2687 | XP_005253615.1 | |
| XM_011538095.2 | 12 | 2970 | XP_011536397.1 | |
| XM_011538096.2 | 12 | 2766 | XP_011536398.1 | |
| XM_011538098.2 | 11 | 3612 | XP_011536400.1 | |
| XM_017019103.1 | 13 | 2808 | XP_016874592.1 | |
| XM_017019104.1 | 10 | 2106 | XP_016874593.1 | |
| AB040953.2 | 12 | 5276 | ||
| AB112582.1 | BAC98409.1 | |||
| AB112583.1 | BAC98410.1 | |||
| AB177852.1 | 12 | 2659 | ||
| AF216494.1 | 12 | 2735 | AAF89993.1 | |
| AK074738.1 | 9 | 1691 | BAC11171.1 | |
| AK304295.1 | 11 | 2554 | ||
| BC064384.1 | 12 | 3024 | AAH64384.1 | |
| U66676.1 | 3 | 582 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv22n68 | Chr.12:122911380 - 123104743 on Build GRCh38 | Loss |
|
| nsv517200 | Chr.12:122827504 - 123169951 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map