Assay Details
Target Gene Details
Entrez Gene ID: | 493856 |
Gene Name: | CDGSH iron sulfur domain 2 |
Gene Aliases: |
ERIS, Miner1, NAF-1, WFS2, ZCD2 |
Location: |
Chr.4:102868978-102892807 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CISD2 | NM_001008388.4 | 2 | 255 | NP_001008389.1 |
| AK292134.1 | 2 | 192 | ||
| BC032300.1 | 2 | 192 | AAH32300.1 | |
| BX537971.1 | 2 | 193 | CAD97935.1 | |
| DN993861.1 | 2 | 260 |
Target Gene Details
Entrez Gene ID: | 150159 |
Gene Name: | solute carrier family 9 member B1 |
Gene Aliases: |
NHA1, NHEDC1 |
Location: |
Chr.4:102885048-103019739 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC9B1 | NM_001100874.2 | 12 | 1543 | NP_001094344.1 |
| NR_047513.1 | 11 | 1415 | ||
| NR_047515.1 | ||||
| AY461581.1 | 12 | 1491 | ||
| DB526003.1 | 2 | 208 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759270 | Chr.4:102859548 - 103136164 on Build GRCh38 | Gain |
|
| nsv594957 | Chr.4:102806853 - 102948934 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map