Assay Details
Target Gene Details
Entrez Gene ID: | 79132 |
Gene Name: | DEXH-box helicase 58 |
Gene Aliases: |
D11LGP2, D11lgp2e, LGP2, RLR-3 |
Location: |
Chr.17:42101404-42112733 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHX58 | NM_024119.2 | 14 | 2199 | NP_077024.2 |
| XM_017025059.1 | 13 | 2132 | XP_016880548.1 | |
| XM_017025060.1 | 14 | 2195 | XP_016880549.1 | |
| AK021416.1 | 14 | 2199 | BAB13818.1 | |
| AK097669.1 | 12 | 1840 | ||
| AK225549.1 | ||||
| AK300900.1 | 14 | 2129 | ||
| BC014949.1 | 14 | 2136 | AAH14949.1 | |
| BM723672.1 | 2 | 194 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056904 | Chr.17:42025823 - 42363204 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map