Assay Details
Target Gene Details
Entrez Gene ID: | 144608 |
Gene Name: | chromosome 12 open reading frame 60 |
Gene Aliases: |
- |
Location: |
Chr.12:14567991-14823858 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C12orf60 | XM_017018871.1 | XP_016874360.1 | ||
| XM_017018872.1 | XP_016874361.1 |
Target Gene Details
Entrez Gene ID: | 55766 |
Gene Name: | H2A histone family member J |
Gene Aliases: |
H2AJ |
Location: |
Chr.12:14774336-14778002 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| H2AFJ | NM_177925.3 | 1 | 2101 | NP_808760.1 |
| NR_027716.2 | 2 | 1787 | ||
| AK001765.1 | 2 | 1547 | BAA91894.1 | |
| AL133626.1 | 2 | 1541 |
More Information
Set Membership: |
|

Genomic Map