Assay Details
Target Gene Details
Entrez Gene ID: | 11098 |
Gene Name: | protease, serine 23 |
Gene Aliases: |
SIG13, SPUVE, ZSIG13 |
Location: |
Chr.11:86791059-86952910 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRSS23 | NM_001293178.1 | 3 | 2511 | NP_001280107.1 |
| NM_001293179.1 | 2 | 2602 | NP_001280108.1 | |
| NM_001293180.1 | 2 | 2332 | NP_001280109.1 | |
| NM_007173.5 | 2 | 2607 | NP_009104.2 | |
| NR_120591.1 | ||||
| NR_120592.1 | ||||
| NR_120593.1 | ||||
| AK075362.1 | 2 | 2303 | ||
| AU279649.1 | ||||
| DA396057.1 | ||||
| DA600625.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3627099 | Chr.11:86660723 - 87045565 on Build GRCh38 | Gain |
|
| nsv523704 | Chr.11:86797886 - 86817395 on Build GRCh38 | Loss |
|
| esv2760199 | Chr.11:86801270 - 86820076 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map